CAS RN 13362-30-6
CAS RN 13362-30-6
IUPAC Name:
2-amino-4-(ethoxycarbonyl)pyridin-1-ium
Synonyms:
2-amino-4-(ethoxycarbonyl)pyridin-1-ium
Molecular Weight (g/mol):
167.19
Molecular Formula:
C8H11N2O2
InChi Key:
XVBZFXZNJAFCHL-UHFFFAOYSA-O
SMILES:
CCOC(=O)C1=CC=[NH+]C(N)=C1
1
–
1
of
1
results
1
Ethyl 2-aminopyridine-4-carboxylate, 97%
CAS: 13362-30-6 Molecular Formula: C8H10N2O2 Molecular Weight (g/mol): 166.18 MDL Number: MFCD03791260 InChI Key: XVBZFXZNJAFCHL-UHFFFAOYSA-N Synonym: ethyl 2-aminoisonicotinate,2-amino-isonicotinic acid ethyl ester,ethyl 2-amino-4-pyridinecarboxylate,2-aminoisonicotinic acid ethyl ester,2-amino-4-ethoxycarbonyl pyridine,4-pyridinecarboxylic acid, 2-amino-, ethyl ester,2-amino-4-pyridinecarboxylic acid ethyl ester,2-aminopyridine-4-carboxylic acid ethyl ester,pubchem16599,acmc-209bt6 PubChem CID: 7015521 IUPAC Name: ethyl 2-aminopyridine-4-carboxylate SMILES: CCOC(=O)C1=CC(=NC=C1)N