accessibility menu, dialog, popup

UserName

CH3SiCH3H3CHC

CAS RN 13361-64-3

IUPAC Name: trimethyl(prop-2-yn-1-yl)silane
Synonyms: trimethyl(prop-2-yn-1-yl)silane
Molecular Weight (g/mol): 112.25
Molecular Formula: C6H12Si
InChi Key: ULYLMHUHFUQKOE-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CC#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)