accessibility menu, dialog, popup

UserName

CAS RN 13331-27-6

OHBHONOO

CAS RN 13331-27-6

IUPAC Name: (3-nitrophenyl)boronic acid
Synonyms: 3-nitrophenylboronic acid
Molecular Weight (g/mol): 166.93
Molecular Formula: C6H6BNO4
InChi Key: ZNRGSYUVFVNSAW-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)