CAS RN 1329611-47-3
CAS RN 1329611-47-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(R)-De(aminosulfonyl) Tamsulosin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(R)-De(aminosulfonyl) Tamsulosin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(R)-De(aminosulfonyl) Tamsulosin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.