CAS RN 1328-53-6
IUPAC Name:
3,4,5,6,12,14,15,21,22,23,24,30,31,32,33-pentadecachloro-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-2,4,6,8,11,13,15,17(41),18,20,22,24,26,28(40),29,31,33,35-octadecaene
Synonyms:
3,4,5,6,12,14,15,21,22,23,24,30,31,32,33-pentadecachloro-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-2,4,6,8,11,13,15,17(41),18,20,22,24,26,28(40),29,31,33,35-octadecaene polychloro copper phthalocyanine (pigment green 7
Molecular Weight (g/mol):
1094.73
Molecular Formula:
C32H3Cl15CuN8
InChi Key:
BWWFBQOKGNVMQO-UHFFFAOYNA-N
SMILES:
ClC1=C2C3N=C(N=C4N5[Cu]N6C(N=C7N=C(N=C5C5=C(Cl)C(Cl)=C(Cl)C(Cl)=C45)C4=C(Cl)C(Cl)=C(Cl)C(Cl)=C74)C4=C(Cl)C(Cl)=C(Cl)C(Cl)=C4C6=N3)C2=C(Cl)C(Cl)=C1
1
–
1
of
1
results
1
Pigment Green 7 (Technical Grade), TRC
CAS: 1328-53-6 Molecular Formula: C32Cl16CuN8 Molecular Weight (g/mol): 1127.19 SMILES: ClC1=C(Cl)C(Cl)=C(Cl)C2=C3/N=C\4C5=C(Cl)C(Cl)=C(Cl)C(Cl)=C5C(=N4)N=C6C7=C(Cl)C(Cl)=C(Cl)C(Cl)=C7C8=NC(=N9)C%10=C(Cl)C(Cl)=C(Cl)C(Cl)=C%10C9=NC(=C12)N3[Cu]N68