accessibility menu, dialog, popup

UserName

HNNHHBrHBr

CAS RN 132747-20-7

IUPAC Name: 2,5-diazabicyclo[2.2.1]heptane dihydrobromide
Synonyms: 2,5-diazabicyclo[2.2.1]heptane dihydrobromide
Molecular Weight (g/mol): 259.97
Molecular Formula: C5H12Br2N2
InChi Key: ISYQWKOXKGJREA-UHFFFAOYNA-N
SMILES: Br.Br.C1NC2CNC1C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results