accessibility menu, dialog, popup

UserName

CAS RN 132705-51-2

NPNNBrFPFFFFF

CAS RN 132705-51-2

IUPAC Name: bromotris(pyrrolidin-1-yl)phosphanium; hexafluoro-λ⁵-phosphanuide
Synonyms: bromotris(pyrrolidin-1-yl)phosphanium hexafluorophosphate
Molecular Weight (g/mol): 466.19
Molecular Formula: C12H24BrF6N3P2
InChi Key: CYKRMWNZYOIJCH-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.Br[P+](N1CCCC1)(N1CCCC1)N1CCCC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)