accessibility menu, dialog, popup

UserName

CH3OFOHCl

CAS RN 1323955-60-7

IUPAC Name: (4-chloro-3-ethoxy-2-fluorophenyl)methanol
Synonyms: (4-chloro-3-ethoxy-2-fluorophenyl)methanol
Molecular Weight (g/mol): 204.63
Molecular Formula: C9H10ClFO2
InChi Key: KEZBCIIBPSEUHP-UHFFFAOYSA-N
SMILES: CCOC1=C(Cl)C=CC(CO)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results