accessibility menu, dialog, popup

UserName

CAS RN 13224-84-5

CH3SiCH3CH3HO

CAS RN 13224-84-5

IUPAC Name: 5-(trimethylsilyl)pent-4-yn-1-ol
Synonyms: 5-(trimethylsilyl)pent-4-yn-1-ol
Molecular Weight (g/mol): 156.3
Molecular Formula: C8H16OSi
InChi Key: LIHAMNSEAAPIRM-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CCCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results