accessibility menu, dialog, popup

UserName

CH3NHO

CAS RN 13220-33-2

IUPAC Name: 1-methylpyrrolidin-3-ol
Synonyms: 1-methylpyrrolidin-3-ol
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: FLVFPAIGVBQGET-UHFFFAOYNA-N
SMILES: CN1CCC(O)C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)