CAS RN 1315629-79-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Atorvastatin Cyclic (Fluorophenyl) Sodium Salt Impurity, TRC
CAS: 1315629-79-8 Molecular Formula: C33 H34 F N2 O7 . Na Molecular Weight (g/mol): 612.62 Synonym: 1b-(4-Fluorophenyl)hexahydro-β,7-dihydroxy-7-(1-methylethyl)-1a-phenyl-7a-[(phenylamino)carbonyl]-3H-oxireno[3,4]pyrrolo[2,1-b][1,3]oxazine-3-butanoic Acid Sodium Salt (1:1) IUPAC Name: sodium;4-[1b-(4-fluorophenyl)-7-hydroxy-1a-phenyl-7a-(phenylcarbamoyl)-7-propan-2-yl-4,5-dihydro-3H-oxireno[1,2]pyrrolo[3,5-b][1,3]oxazin-3-yl]-3-hydroxybutanoate SMILES: [Na+].CC(C)C1(O)N2CCC(CC(O)CC(=O)[O-])OC2(c3ccc(F)cc3)C4(OC14C(=O)Nc5ccccc5)c6ccccc6