accessibility menu, dialog, popup

UserName

OWOO

CAS RN 1314-35-8

IUPAC Name: trioxotungsten
Synonyms: tungsten trioxide
Molecular Weight (g/mol): 231.84
Molecular Formula: O3W
InChi Key: ZNOKGRXACCSDPY-UHFFFAOYSA-N
SMILES: O=[W](=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
  • (1)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (7)
  • (1)
  • (1)