CAS RN 131275-93-9
CAS RN 131275-93-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(3R,5S)-Atorvastatin Sodium Salt, TRC
CAS: 131275-93-9 Molecular Formula: C33 H34 F N2 O5 . Na Molecular Weight (g/mol): 580.62 Synonym: 1H-Pyrrole-1-heptanoic acid, 2-(4-fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-, monosodium salt, (R*,S*)- (9CI),(3RS,5SR)-7-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]-3,5-dihydroxyheptanoic acid sodium salt,Atorvastatin Calcium Trihydrate Imp. B (EP) as Sodium Salt IUPAC Name: sodium;(3R,5S)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate SMILES: [Na+].CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@H](O)C[C@@H](O)CC(=O)[O-]