accessibility menu, dialog, popup

UserName

OOOInIn

CAS RN 1312-43-2

IUPAC Name: diindium(3+) trioxidandiide
Synonyms: indium(III) oxide
Molecular Weight (g/mol): 277.63
Molecular Formula: In2O3
InChi Key: PJXISJQVUVHSOJ-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[In+3].[In+3]
Quantity 
  • (8)
  • (1)
  • (2)
  • (3)
  • (2)
  • (6)
Physical Form 
  • (1)
  • (2)
  • (13)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (2)
Grade 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (19)
  • (2)
  • (8)
  • (1)
  • (2)
  • (3)
  • (2)
  • (6)
  • (1)
  • (2)
  • (13)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)