accessibility menu, dialog, popup

UserName

CAS RN 1312-41-0

InSb

CAS RN 1312-41-0

IUPAC Name: indium(3+) antimony
Synonyms: indium antimonide
Molecular Weight (g/mol): 236.58
Molecular Formula: InSb
InChi Key: BGRWVDLWYMJUQP-UHFFFAOYSA-N
SMILES: [In+3].[Sb]
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (3)