accessibility menu, dialog, popup

UserName

CAS RN 131-53-3

H3COOHOOH

CAS RN 131-53-3

IUPAC Name: 2-(2-hydroxybenzoyl)-5-methoxyphenol
Synonyms: dioxybenzone dioxybenzone (substance) dioxibenzona dioxibenzonum cyasorb UV 24 dioxybenzon spectra-sorb UV 24 dioxybenzone preparation dioxybenzonum benzophenone-8
Molecular Weight (g/mol): 244.25
Molecular Formula: C14H12O4
InChi Key: MEZZCSHVIGVWFI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(=O)C2=CC=CC=C2O)C(O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results