accessibility menu, dialog, popup

UserName

CAS RN 131-16-8

H3COOOOCH3

CAS RN 131-16-8

IUPAC Name: 1,2-dipropyl benzene-1,2-dicarboxylate
Synonyms: dipropyl phthalate di-n-propylphthalate N-dipropyl phthalate
Molecular Weight (g/mol): 250.29
Molecular Formula: C14H18O4
InChi Key: MQHNKCZKNAJROC-UHFFFAOYSA-N
SMILES: CCCOC(=O)C1=CC=CC=C1C(=O)OCCC
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (7)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)