accessibility menu, dialog, popup

UserName

OHOHMg

CAS RN 1309-42-8

IUPAC Name: magnesium(2+) dihydroxide
Synonyms: magnesia, magnesiumhydroxid magnesium hydroxide magnesium hydrate Mg(OH)2 milk of magnesia
Molecular Weight (g/mol): 58.32
Molecular Formula: H2MgO2
InChi Key: VTHJTEIRLNZDEV-UHFFFAOYSA-L
SMILES: [OH-].[OH-].[Mg++]
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
Physical Form 
  • (6)
  • (2)
Grade 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (6)
  • (2)
  • (3)
  • (3)