accessibility menu, dialog, popup

UserName

FeFeOOO

CAS RN 1309-37-1

IUPAC Name: diiron(3+) hydrate trioxidandiide
Synonyms: iron(III) oxide
Molecular Weight (g/mol): 177.7
Molecular Formula: Fe2H2O4
InChi Key: QGPQTSCLUYMZHL-UHFFFAOYSA-N
SMILES: O.[O--].[O--].[O--].[Fe+3].[Fe+3]
Quantity 
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (7)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (17)
  • (2)
  • (2)
Percent Purity 
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
Grade 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (6)
  • (1)
  • (4)
  • (13)
  • (3)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (7)
  • (1)
  • (1)
  • (2)
  • (17)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)