accessibility menu, dialog, popup

UserName

CAS RN 13073-25-1

OHBrNOO

CAS RN 13073-25-1

IUPAC Name: 2-bromo-6-nitrophenol
Synonyms: 2-bromo-6-nitrophenol
Molecular Weight (g/mol): 218.01
Molecular Formula: C6H4BrNO3
InChi Key: VEJSIOPQKQXJAT-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)