accessibility menu, dialog, popup

UserName

CAS RN 1306761-53-4

H3CCH3ONNOHNOOHNHCl

CAS RN 1306761-53-4

IUPAC Name: 3-[(4-{5-[3-cyano-4-(propan-2-yloxy)phenyl]-1,2,4-oxadiazol-3-yl}-2,3-dihydro-1H-inden-1-yl)amino]propanoic acid hydrochloride
Synonyms: 3-({4-[5-(3-cyano-4-isopropoxyphenyl)-1,2,4-oxadiazol-3-yl]-2,3-dihydro-1H-inden-1-yl}amino)propanoic acid hydrochloride
Molecular Weight (g/mol): 468.94
Molecular Formula: C24H25ClN4O4
InChi Key: ZHPGSGYEFGEZAL-UHFFFAOYNA-N
SMILES: Cl.CC(C)OC1=C(C=C(C=C1)C1=NC(=NO1)C1=C2CCC(NCCC(O)=O)C2=CC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results