accessibility menu, dialog, popup

UserName

CAS RN 130628-75-0

CH3NOBr

CAS RN 130628-75-0

IUPAC Name: 3-(bromomethyl)-5-methyl-1,2-oxazole
Synonyms: 3-(bromomethyl)-5-methyl-1,2-oxazole
Molecular Weight (g/mol): 176.01
Molecular Formula: C5H6BrNO
InChi Key: ASGJFGPILHALRC-UHFFFAOYSA-N
SMILES: CC1=CC(CBr)=NO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results