accessibility menu, dialog, popup

UserName

CAS RN 13057-17-5

CH3OBr

CAS RN 13057-17-5

IUPAC Name: bromo(methoxy)methane
Synonyms: methane, bromomethoxy-
Molecular Weight (g/mol): 124.97
Molecular Formula: C2H5BrO
InChi Key: JAMFGQBENKSWOF-UHFFFAOYSA-N
SMILES: COCBr
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)