accessibility menu, dialog, popup

UserName

SH2NClF

CAS RN 130560-97-3

IUPAC Name: 3-chloro-4-fluorobenzene-1-carbothioamide
Synonyms: 3-chloro-4-fluorobenzenecarbothioamide
Molecular Weight (g/mol): 189.63
Molecular Formula: C7H5ClFNS
InChi Key: RSGPCKCTKDCMRR-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC(Cl)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results