accessibility menu, dialog, popup

UserName

CAS RN 13055-63-5

CH3OHOFFF

CAS RN 13055-63-5

IUPAC Name: 2-methyl-5-(trifluoromethyl)benzoic acid
Synonyms: 2-methyl-5-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: STJUSBSLJXOSDU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1C(O)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results