accessibility menu, dialog, popup

UserName

CAS RN 1304-29-6

BaOO

CAS RN 1304-29-6

IUPAC Name: barium(2+) dioxidanediide
Synonyms: barium peroxide
Molecular Weight (g/mol): 169.33
Molecular Formula: BaO2
InChi Key: ZJRXSAYFZMGQFP-UHFFFAOYSA-N
SMILES: [Ba++].[O-][O-]
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (8)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (6)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (8)
  • (1)
  • (1)
  • (1)
  • (3)