accessibility menu, dialog, popup

UserName

CAS RN 13031-43-1

IUPAC Name: 4-acetylphenyl acetate
Synonyms: P-acetoxyacetophenone
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: SMIOEQSLJNNKQF-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(C=C1)C(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)