accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHOHO

CAS RN 130309-46-5

IUPAC Name: 4-{[(tert-butoxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Synonyms: 4-[(tert-butoxycarbonyl)amino]cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 243.3
Molecular Formula: C12H21NO4
InChi Key: KXMRDHPZQHAXML-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(CC1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)