accessibility menu, dialog, popup

UserName

H3CNCH3CH3CH3H3C

CAS RN 13021-15-3

IUPAC Name: N,N,2,4,6-pentamethylaniline
Synonyms: N,N,2,4,6-pentamethylaniline
Molecular Weight (g/mol): 163.26
Molecular Formula: C11H17N
InChi Key: JZBZLRKFJWQZHU-UHFFFAOYSA-N
SMILES: CN(C)C1=C(C)C=C(C)C=C1C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results