accessibility menu, dialog, popup

UserName

NNH2NNH2HClHClHClHCl

CAS RN 1301706-42-2

IUPAC Name: 1,2-bis(pyridin-3-yl)ethane-1,2-diamine tetrahydrochloride
Synonyms: 1,2-bis(pyridin-3-yl)ethane-1,2-diamine tetrahydrochloride
Molecular Weight (g/mol): 360.1
Molecular Formula: C12H18Cl4N4
InChi Key: GAHCWAYKHBFBHN-UHFFFAOYNA-N
SMILES: Cl.Cl.Cl.Cl.NC(C(N)C1=CN=CC=C1)C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results