CAS RN 1300031-49-5
CAS RN 1300031-49-5
IUPAC Name:
7-(3,5-dimethyl-1,2-oxazol-4-yl)-8-methoxy-1-[1-(pyridin-2-yl)ethyl]-1H,2H,3H-imidazo[4,5-c]quinolin-2-one
Synonyms:
7-(3,5-dimethyl-1,2-oxazol-4-yl)-8-methoxy-1-[1-(pyridin-2-yl)ethyl]-3H-imidazo[4,5-c]quinolin-2-one
Molecular Weight (g/mol):
415.45
Molecular Formula:
C23H21N5O3
InChi Key:
VUVUVNZRUGEAHB-UHFFFAOYNA-N
SMILES:
COC1=C(C=C2N=CC3=C(N(C(C)C4=CC=CC=N4)C(=O)N3)C2=C1)C1=C(C)ON=C1C
1
–
3
of
3
results
2
3
I-BET151, MedChemExpress
MedChemExpress I-BET151 (GSK1210151A) is a BET bromodomain inhibitor which inhibits BRD4, BRD2, and BRD3 with pIC50 of 6.1, 6.3, and 6.6, respectively.