accessibility menu, dialog, popup

UserName

NaOHHOOOSOOO

CAS RN 130-22-3

IUPAC Name: sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
Synonyms: sodium alizarinesulfonate
Molecular Weight (g/mol): 342.25
Molecular Formula: C14H7NaO7S
InChi Key: HFVAFDPGUJEFBQ-UHFFFAOYSA-M
SMILES: [Na+].OC1=C(O)C2=C(C=C1S([O-])(=O)=O)C(=O)C1=CC=CC=C1C2=O
Quantity 
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (9)
  • (1)
Color 
  • (3)
  • (3)
  • (2)
Grade 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (4)
  • (5)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (9)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)