accessibility menu, dialog, popup

UserName

Sm

CAS RN 1298-55-1

IUPAC Name: tris(cyclopenta-2,4-dien-1-yl)samarium
Synonyms: tris(cyclopenta-2,4-dien-1-yl)samarium
Molecular Weight (g/mol): 345.65
Molecular Formula: C15H15Sm
InChi Key: HLLVKWMULLMCMH-UHFFFAOYSA-N
SMILES: C1=CC(C=C1)[Sm](C1C=CC=C1)C1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results