accessibility menu, dialog, popup

UserName

CH3OO(S)NH2NOOCH3H3CCH3

CAS RN 129799-08-2

IUPAC Name: (2S)-4-[(tert-butoxy)carbonyl]-2-(methoxycarbonyl)piperazin-1-ium
Synonyms: (2S)-4-(tert-butoxycarbonyl)-2-(methoxycarbonyl)piperazin-1-ium
Molecular Weight (g/mol): 245.3
Molecular Formula: C11H21N2O4
InChi Key: QUKAHFCVKNRRBU-QMMMGPOBSA-O
SMILES: COC(=O)[C@@H]1CN(CC[NH2+]1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results