accessibility menu, dialog, popup

UserName

CAS RN 129535-78-0

CH3HNO

CAS RN 129535-78-0

IUPAC Name: methyl[(3-phenoxyphenyl)methyl]amine
Synonyms: methyl[(3-phenoxyphenyl)methyl]amine
Molecular Weight (g/mol): 213.28
Molecular Formula: C14H15NO
InChi Key: GDVLOOQWWBPGSV-UHFFFAOYSA-N
SMILES: CNCC1=CC(OC2=CC=CC=C2)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results