accessibility menu, dialog, popup

UserName

CAS RN 128-95-0

NH2NH2OO

CAS RN 128-95-0

IUPAC Name: 1,4-diamino-9,10-dihydroanthracene-9,10-dione
Synonyms: disperse violet 1
Molecular Weight (g/mol): 238.25
Molecular Formula: C14H10N2O2
InChi Key: FBMQNRKSAWNXBT-UHFFFAOYSA-N
SMILES: NC1=CC=C(N)C2=C1C(=O)C1=CC=CC=C1C2=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results