CAS RN 127780-16-9
CAS RN 127780-16-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
5-(Difluoromethoxy)-2-[[(3,4-dimethoxypyridin-2-yl)methyl]sulphonyl]-1H-benzimidazole (Pantoprazole Sulphone), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Pantoprazole Sulfone, TRC
CAS: 127780-16-9 Molecular Formula: C16 H15 F2 N3 O5 S Molecular Weight (g/mol): 399.37 Synonym: 5-(Difluoromethoxy)-2-[[(3,4-dimethoxypyridin-2-yl)methyl]sulphonyl]-1H-benzimidazole,Pantoprazole Sodium Sesquihydrate Imp. A (EP),Pantoprazole Sulphone IUPAC Name: 5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methylsulfonyl]-1H-benzimidazole SMILES: COc1ccnc(CS(=O)(=O)c2nc3cc(OC(F)F)ccc3[nH]2)c1OC