accessibility menu, dialog, popup

UserName

CoHCHC

CAS RN 1277-43-6

IUPAC Name: λ²-cobalt(2+) bis(cyclopenta-2,4-dien-1-ide)
Synonyms: cobaltocene
Molecular Weight (g/mol): 189.12
Molecular Formula: C10H10Co
InChi Key: ILZSSCVGGYJLOG-UHFFFAOYSA-N
SMILES: [Co++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)