accessibility menu, dialog, popup

UserName

CAS RN 127406-55-7

ON

CAS RN 127406-55-7

IUPAC Name: 4-(pyridin-3-yl)benzaldehyde
Synonyms: 4-(pyridin-3-yl)benzaldehyde
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: NXZUVHZZIZHEOP-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results