accessibility menu, dialog, popup

UserName

CAS RN 1272-23-7

CCCLa

CAS RN 1272-23-7

IUPAC Name: lanthanum(3+) tris(cyclopenta-1,3-dien-1-ide)
Synonyms: lanthanum(3+) tris(cyclopenta-1,3-dien-1-ide)
Molecular Weight (g/mol): 334.19
Molecular Formula: C15H15La
InChi Key: WMTALVRZNMWNTJ-UHFFFAOYSA-N
SMILES: [La+3].C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results