CAS RN 127053-22-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(E)-4-(6-Methoxynaphthalen-2-yl)but-3-en-2-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Dehydro Nabumetone, TRC
CAS: 127053-22-9 Molecular Formula: C15 H14 O2 Molecular Weight (g/mol): 226.27 Synonym: (E)-4-(6-Methoxynaphthalen-2-yl)but-3-en-2-one,Nabumetone Imp. D (EP),3-Buten-2-one, 4-(6-methoxy-2-naphthalenyl)-, (E)- (9CI),(3E)-4-(6-Methoxy-2-naphthalenyl)-3-buten-2-one,trans-1-(2'-Methoxynaphth-6'-yl)but-1-en-3-one,(E)-4-(6-Methoxy-2-naphthyl)-3-buten-2-one IUPAC Name: (E)-4-(6-methoxynaphthalen-2-yl)but-3-en-2-one SMILES: COc1ccc2cc(\C=C\C(=O)C)ccc2c1