accessibility menu, dialog, popup

UserName

H3COHCH

CAS RN 127-66-2

IUPAC Name: 2-phenylbut-3-yn-2-ol
Synonyms: 2-phenylbut-3-yn-2-ol
Molecular Weight (g/mol): 146.19
Molecular Formula: C10H10O
InChi Key: KSLSOBUAIFEGLT-UHFFFAOYNA-N
SMILES: CC(O)(C#C)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results