accessibility menu, dialog, popup

UserName

CAS RN 126937-41-5

CH3H3CCH3OONNOOHOO

CAS RN 126937-41-5

IUPAC Name: 1-[(benzyloxy)carbonyl]-4-[(tert-butoxy)carbonyl]piperazine-2-carboxylic acid
Synonyms: 1-[(benzyloxy)carbonyl]-4-(tert-butoxycarbonyl)piperazine-2-carboxylic acid
Molecular Weight (g/mol): 364.4
Molecular Formula: C18H24N2O6
InChi Key: MKXMXZZARNRMMQ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCN(C(C1)C(O)=O)C(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results