CAS RN 126924-38-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-Norfluoxetine, TRC
CAS: 126924-38-7 Molecular Formula: C16 H16 F3 N O Molecular Weight (g/mol): 295.3 Synonym: Benzenepropanamine, γ-[4-(trifluoromethyl)phenoxy]-, (γS)- (9CI, ACI),(γS)-γ-[4-(Trifluoromethyl)phenoxy]benzenepropanamine (ACI),Benzenepropanamine, γ-[4-(trifluoromethyl)phenoxy]-, (S)- (ZCI),(3S)-3-Phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine,(S)-3-(4-Trifluoromethylphenoxy)-3-phenylpropylamine,(S)-Norfluoxetine,1-[(1S)-3-Amino-1-phenylpropoxy]-4-(trifluoromethyl)benzene,Seproxetine IUPAC Name: (3S)-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine SMILES: NCC[C@H](OC1=CC=C(C(F)(F)F)C=C1)C2=CC=CC=C2