accessibility menu, dialog, popup

UserName

FH2NFBr

CAS RN 1262198-07-1

IUPAC Name: 3-bromo-2,6-difluoroaniline
Synonyms: 3-bromo-2,6-difluoroaniline
Molecular Weight (g/mol): 208.01
Molecular Formula: C6H4BrF2N
InChi Key: HIZADWSSPWORHH-UHFFFAOYSA-N
SMILES: NC1=C(F)C=CC(Br)=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results