accessibility menu, dialog, popup

UserName

CAS RN 126162-38-7

IUPAC Name: 2,3-difluoro-4-hydroxybenzonitrile
Synonyms: 2,3-difluoro-4-hydroxybenzonitrile
Molecular Weight (g/mol): 155.1
Molecular Formula: C7H3F2NO
InChi Key: UIJJJWADIVZXNT-UHFFFAOYSA-N
SMILES: OC1=C(F)C(F)=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results