accessibility menu, dialog, popup

UserName

CAS RN 1261445-13-9

CH3OFFFOCl

CAS RN 1261445-13-9

IUPAC Name: 2-methyl-5-(trifluoromethoxy)benzoyl chloride
Synonyms: 2-methyl-5-(trifluoromethoxy)benzoyl chloride
Molecular Weight (g/mol): 238.59
Molecular Formula: C9H6ClF3O2
InChi Key: DWHYIYNVPCGCJQ-UHFFFAOYSA-N
SMILES: CC1=C(C=C(OC(F)(F)F)C=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results