accessibility menu, dialog, popup

UserName

CAS RN 126-30-7

CH3H3COHHO

CAS RN 126-30-7

IUPAC Name: 2,2-dimethylpropane-1,3-diol
Synonyms: neopentyl glycol
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: SLCVBVWXLSEKPL-UHFFFAOYSA-N
SMILES: CC(C)(CO)CO
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)