accessibility menu, dialog, popup

UserName

CAS RN 1256486-24-4

NHONH2NHNNOHNH2(E/Z)(E/Z)

CAS RN 1256486-24-4

IUPAC Name: N'-hydroxy-2-[3-(N'-hydroxycarbamimidoyl)phenyl]-1H-1,3-benzodiazole-6-carboximidamide
Synonyms: N'-hydroxy-2-[3-(N'-hydroxycarbamimidoyl)phenyl]-3H-1,3-benzodiazole-5-carboximidamide
Molecular Weight (g/mol): 310.32
Molecular Formula: C15H14N6O2
InChi Key: KXTHKUXQYDDRHI-UHFFFAOYSA-N
SMILES: NC(=NO)C1=CC=C2N=C(NC2=C1)C1=CC(=CC=C1)C(N)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results