CAS RN 125481-61-0
CAS RN 125481-61-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6’-Hydroxy Buspirone, TRC
CAS: 125481-61-0 Molecular Formula: C21 H31 N5 O3 Molecular Weight (g/mol): 401.5 Synonym: 8-Azaspiro[4.5]decane-7,9-dione, 6-hydroxy-8-[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]-,6-Hydroxy-8-[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]-8-azaspiro[4.5]decane-7,9-dione,6-Hydroxybuspirone,6'-Hydroxybuspirone,BMS 528215,BMY 28674 IUPAC Name: 10-hydroxy-8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione SMILES: OC1C(=O)N(CCCCN2CCN(CC2)c3ncccn3)C(=O)CC14CCCC4